missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium, plasma standard solution, Specpure™ Sr 1000μg/mL, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 013874AE

 View more versions of this product

Catalog No. AA13874AE



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Strontium, plasma standard solution
100 mL
MFCD00134060
23 hPa (17mm Hg) at 20°C
Sr 1000μg/mL
Sr(NO3)2 in 5% HNO3
Nitric acid: 5%; Strontium nitrate: 0.24%; Water: 94.76%
Liquid

Chemical Identifiers

Sr(NO3)2 in 5% HNO3
MFCD00134060
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : DOT Class: 8; Packaging Group: III

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only