missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium oxide, tech.

Manufacturer:  Alfa Aesar™ 1249530

 View more versions of this product

Catalog No. AA1249530



Description

Description

  • In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant
Specifications

Specifications

Strontium oxide
OSr
UN3262
Soluble in fused potassium hydroxide; Insoluble in ether and acetone; Decomposes in water to Sr(OH)2 with evolution of heat
O=[Sr]
103.619
103.62
Technical
−100 Mesh
Moisture Sensitive
2430°C
1314-11-0
MFCD00049558
14,8846
UFQXGXDIJMBKTC-UHFFFAOYSA-N
oxostrontium
518709
Powder
4.700g/mL
Odorless
~3000°C
250g

Chemical Identifiers

1314-11-0
103.619
UFQXGXDIJMBKTC-UHFFFAOYSA-N
oxostrontium
OSr
MFCD00049558
518709
O=[Sr]
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents