missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium oxide, 99.5% (metals basis), SrO ≈97%, Alfa Aesar™

Manufacturer:  Alfa Aesar 8822014

 View more versions of this product

Catalog No. AA8822014



Specifications

Specifications

Strontium oxide
99.5% (metals basis)
MFCD00049558
O=[Sr]
103.619
103.62
4.7g/mL
1314-11-0
OSr
UFQXGXDIJMBKTC-UHFFFAOYSA-N
oxostrontium
518709
-100 Mesh Powder
25g

Chemical Identifiers

1314-11-0
103.619
UFQXGXDIJMBKTC-UHFFFAOYSA-N
oxostrontium
OSr
MFCD00049558
518709
O=[Sr]
Safety and Handling

Safety and Handling

EINECSNumber : 215-219-9

SDS Documents