Please login to your online account to display your discounted pricing

Strontium oxide, 99.5% (metals basis), SrO ≈97%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Strontium oxide
CAS 1314-11-0
Physical Form -100 Mesh Powder
Assay Percent Range 99.5% (metals basis)
Molecular Formula SrO
Formula Weight 103.62
MDL Number MFCD00049558
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8822006

 
alfa aesar
8822006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8822014

 
alfa aesar
8822014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8822022

 
alfa aesar
8822022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form -100 Mesh Powder
Formula Weight 103.62
Quantity 5g
Chemical Name or Material Strontium oxide
EINECS Number 215-219-9
Assay Percent Range 99.5% (metals basis)
Molecular Formula SrO
MDL Number MFCD00049558
CAS 1314-11-0
Density 4.7g/mL