Learn More

Strontium Nitrate Anhydrous (Crystalline/Certified ACS), Fisher Chemical™
Manufacturer: Fisher Chemical S549500
Specifications
Strontium Nitrate Anhydrous | |
White | |
5.0 to 7.0 | |
≥99 % | |
Sr(NO3)2 | |
bdg873aqzl, hsdb 787, nitrate de strontium french, nitric acid, strontium salt, nitric acid, strontium salt 2:1, strontium dinitrate, strontium ii nitrate 1:2, strontium nitrate, strontium nitrate sr no3 2, unii-bdg873aqzl | |
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
211.63 | |
211.63 | |
≥99% | |
5ppm max. | |
0.01% max. | |
0.1% max. | |
Glass Bottle | |
0.005% max. | |
0.05% max. |
10042-76-9 | |
570°C | |
500g | |
N2O6Sr | |
MFCD00011248 | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
strontium(2+) dinitrate | |
24848 | |
Solid | |
Certified ACS | |
Pass Test | |
5ppm max. | |
0.1% max. | |
0.1% max. | |
0.05% max. | |
0.002% max. |
Chemical Identifiers
10042-76-9 | |
211.63 | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
24848 | |
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
N2O6Sr | |
MFCD00011248 | |
bdg873aqzl, hsdb 787, nitrate de strontium french, nitric acid, strontium salt, nitric acid, strontium salt 2:1, strontium dinitrate, strontium ii nitrate 1:2, strontium nitrate, strontium nitrate sr no3 2, unii-bdg873aqzl | |
strontium(2+) dinitrate |
Safety and Handling

DANGER!
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Irritating to eyes, respiratory system and skin. Use personal protective equipment. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Obtain medical attention. Obtain medical attention. Do not induce vomiting.
NFPA
Health:2
Flammability:0
Instability:2
DOTInformation : DOT Class 5.1, : Oxidizer