missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium nitrate, anhydrous, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 30534D9

 View more versions of this product

Catalog No. AA30534D9



Specifications

Specifications

Anhydrous
98%
MFCD00011248
DHEQXMRUPNDRPG-UHFFFAOYSA-N
strontium;dinitrate
24848
Crystalline or Powder
White
10042-76-9
N2O6Sr
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]
211.628
211.63
Odorless
5 x 1kg

Chemical Identifiers

10042-76-9
211.628
DHEQXMRUPNDRPG-UHFFFAOYSA-N
24848
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]
N2O6Sr
MFCD00011248
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
strontium;dinitrate
SDS Documents