missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium Nitrate ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 10042-76-9
Physical Form Granular
Molecular Formula Sr(NO3)2
Formula Weight 211.63
Chemical Name or Material Strontium Nitrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK8256212 Avantor Performance
825606
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK8256500 Avantor Performance
825604
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10042-76-9
Sr(NO3)2
Strontium Nitrate
Granular
211.63
ACS Grade

Specifications

10042-76-9
Sr(NO3)2
Poly
Strontium Nitrate
Granular
211.63
2.5kg
ACS Grade