missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium nitrate, ACS, 99.0% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1223214

 View more versions of this product

Catalog No. AA1223214



Specifications

Specifications

Strontium nitrate
10042-76-9
N2O6Sr
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]
211.628
211.63
25g
For analysis ACS
99.0% min.
MFCD00011248
DHEQXMRUPNDRPG-UHFFFAOYSA-N
strontium;dinitrate
24848
Crystalline

Chemical Identifiers

10042-76-9
211.628
DHEQXMRUPNDRPG-UHFFFAOYSA-N
24848
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]
N2O6Sr
MFCD00011248
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
strontium;dinitrate
Safety and Handling

Safety and Handling

EINECSNumber : 233-131-9

Documents