missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium nitrate, 99.9965% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1087922

 View more versions of this product

Catalog No. AA1087922



Specifications

Specifications

Strontium nitrate
99.9965% (metals basis)
MFCD00011248
DHEQXMRUPNDRPG-UHFFFAOYSA-N
strontium;dinitrate
24848
Powder, Packaged Under Argon
10042-76-9
N2O6Sr
bdg873aqzl
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]
211.628
211.63
100g

Chemical Identifiers

10042-76-9
211.628
DHEQXMRUPNDRPG-UHFFFAOYSA-N
24848
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]
N2O6Sr
MFCD00011248
bdg873aqzl
strontium;dinitrate
Safety and Handling

Safety and Handling

EINECSNumber : 233-131-9

Documents