Learn More
Strontium nitrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 1087914
Description
Strontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. It mixed with glycolic acid and used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Strontium nitrate | |
10042-76-9 | |
25 g | |
N2O6Sr | |
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1 | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
strontium(2+) dinitrate | |
24848 | |
Hygroscopic |
570°C | |
Powder | |
99.9965% (metals basis) | |
MFCD00011248 | |
Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid. | |
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
211.63 | |
211.63 | |
2.99 g/mL |
Chemical Identifiers
10042-76-9 | |
211.63 | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
24848 |
N2O6Sr | |
MFCD00011248 | |
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1 | |
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
P210-P220-P221-P280-P305+P351+P338-P310-P370+P378q-P501c
H272-H318
DOTInformation : Hazard Class: 5.1; Packaging Group: III
EINECSNumber : 233-131-9
RTECSNumber : WK9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only