missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Strontium nitrate, 98%, extra pure, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438520050
Specifications
Strontium nitrate | |
10042-76-9 | |
N2O6Sr | |
15,8975 | |
Solubility in water: 660g/L (20°C). Other solubilities: slightly soluble in alcohol and acetone | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2] | |
211.63 | |
211.63 | |
98% | |
10ppm max. | |
Plastic bottle | |
White | |
5 to 7 (at 5% 20°C) |
Extra Pure | |
97.5% min. (Complexometry) | |
Sr(NO3)2 | |
bdg873aqzl, hsdb 787, nitrate de strontium french, nitric acid, strontium salt, nitric acid, strontium salt 2:1, strontium dinitrate, strontium ii nitrate 1:2, strontium nitrate, strontium nitrate sr no3 2, unii-bdg873aqzl | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
strontium;dinitrate | |
24848 | |
Crystals or Crystalline Powder | |
Extra Pure | |
0.5% max. | |
Solubility in water: 660g/L (20°C). Other solubilities: slightly soluble in alcohol and acetone | |
570.0°C | |
5kg |
Chemical Identifiers
10042-76-9 | |
211.63 | |
bdg873aqzl, hsdb 787, nitrate de strontium french, nitric acid, strontium salt, nitric acid, strontium salt 2:1, strontium dinitrate, strontium ii nitrate 1:2, strontium nitrate, strontium nitrate sr no3 2, unii-bdg873aqzl | |
strontium;dinitrate |
N2O6Sr | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
24848 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2] |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
EINECSNumber : 233-131-9