Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium neodecanoate, Sr 16-21%

$35.86 - $35.86

Chemical Identifiers

Chemical Name or Material Strontium neodecanoate
CAS 106705-37-7
Formula Weight 430.1
MDL Number MFCD00070439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3957404

 
alfa aesar™
3957404
2g Each for $35.86

AA3957409

 
alfa aesar™
3957409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3957418

 
alfa aesar™
3957418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 430.1
Chemical Name or Material Strontium neodecanoate
Physical Form Powder
MDL Number MFCD00070439
CAS 106705-37-7
Concentration Sr 16 to 21%
Quantity 2g