Learn More
Strontium molybdenum oxide, 99%
CAS: 13470-04-7 | SrMoO4
Supplier: Thermo Scientific Chemicals 04023830
Description
It is used as an optical material for a small range of special applications, for example, as an optical coating on lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Strontium molybdenum oxide | |
| Ba < 0.3% | |
| 13470-04-7 | |
| 250 g | |
| SrMoO4 | |
| Strontium molybdate | |
| 247.56 |
| (decomposition) | |
| Colorless | |
| Powder | |
| 99.9% (excluding Ba) | |
| MFCD00049550 | |
| Insoluble in water. Soluble in acid | |
| 4.54 |
Chemical Identifiers
| 13470-04-7 | |
| SrMoO4 | |
| MFCD00049550 | |
| Strontium molybdate |
Safety and Handling
EINECSNumber : 236-730-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only