missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium lanthanum sulfide

Chemical Identifiers

CAS 12532-80-8
Molecular Formula La2S4Sr
Linear Formula SrLa2S4
Formula Weight 493.67
MDL Number MFCD00061456
UN Number UN3134
Chemical Name or Material Strontium lanthanum sulfide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1701914 SDS Alfa Aesar™
A1701914
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1701906 SDS Alfa Aesar™
A1701906
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1701922 SDS Alfa Aesar™
A1701922
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

12532-80-8
SrLa2S4
MFCD00061456
Strontium lanthanum sulfide
La2S4Sr
493.67
UN3134

Specifications

493.67
12532-80-8
UN3134
25g
La2S4Sr
SrLa2S4
MFCD00061456
Strontium lanthanum sulfide

Safety and Handling

GHS H Statement
H331-H261-H302-H312
Toxic if inhaled.
In contact with water releases flammable gas.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P231+P232-P261-P280-P304+P340-P405-P501a
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.