Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium iodide, anhydrous, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Strontium iodide
CAS 10476-86-5
Name Note Anhydrous
Formula Weight 341.43
MDL Number MFCD00049557
Merck Index 14,8844
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4726806

  SDS
alfa aesar™
4726806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4726814

  SDS
alfa aesar™
4726814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4726822

  SDS
alfa aesar™
4726822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 341.43
Chemical Name or Material Strontium iodide
TSCA TSCA
Odor Odorless
Name Note Anhydrous
Physical Form Powder
Assay 97%
Sensitivity Light Sensitive & Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00049557
CAS 10476-86-5
Merck Index 14,8844
Color Pale yellow to dark brown
Melting Point 515°C
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H290-H335-H315-H319
May be corrosive to metals.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.550g/mL
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H290-H335-H315-H319
May be corrosive to metals.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.