Please login to your online account to display your discounted pricing

Strontium ICP Standard, 1mL = 10mg Sr (10,000ppm Sr), Ricca Chemical


Quantity 500mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIPSR10KN500

 View more versions of this product

Catalog No. PSR10KN-500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Chemical Name or Material Strontium ICP Standard
CAS 7732-18-5
CAS 7697-37-2
Concentration or Composition Notes 10,000ppm Sr
Equivalence 1 mL = 10 mg Sr
CAS Max % 93.0
Packaging Poly natural
CAS Min % 6.7
CAS Min % 89.0
CAS Max % 6.97
CAS Max % 2.24
CAS 10042-76-9
CAS Min % 2.15

Specifications

Chemical Name or Material Strontium ICP Standard
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 7697-37-2
CAS Min % 6.7
CAS Max % 6.97
CAS 10042-76-9
CAS Min % 2.15
CAS Max % 2.24
Physical Form Liquid