missing translation for 'onlineSavingsMsg'
Learn More

Strontium ICP Standard, 1000 ppm Sr in 3% HNO3, Ricca Chemical

$275.50 - $275.50

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PSR1KN-100
View Documents
Ricca Chemical Company
PSR1KN100
100 mL Natural LDPE Bottle
Each for $275.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10042-76-9
211.63
DHEQXMRUPNDRPG-UHFFFAOYSA-N
strontium(2+) dinitrate
N2O6Sr
MFCD00011248
24848
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Natural LDPE Bottle
1,000ppm Sr
97.0,4.39,0.24
Liquid
MFCD00011248
< 2
Standard
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
211.63
24848
10042-76-9,7697-37-2,7732-18-5
94.0,4.22,0.23
1 mL = 1 mg Sr
N2O6Sr
Colorless
Approximately 0°C
DHEQXMRUPNDRPG-UHFFFAOYSA-N
strontium(2+) dinitrate
100 mL
Strontium ICP Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'