Learn More
Strontium hydroxide octahydrate, 99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04135122
Description
Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Strontium hydroxide octahydrate | |
1311-10-0 | |
100 g | |
H18O10Sr | |
Soluble in hot water and acid. Insoluble in acetone. | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++] | |
265.75 | |
Air sensitive; Hygroscopic |
100°C-8H2O | |
Crystalline | |
99% (metals basis) | |
MFCD00150395 | |
UJPWWRPNIRRCPJ-UHFFFAOYSA-L | |
strontium(2+) octahydrate dihydroxide | |
265.76 (121.60 Anhydrous) | |
1.900 g/mL |
Chemical Identifiers
1311-10-0 | |
265.75 | |
UJPWWRPNIRRCPJ-UHFFFAOYSA-L | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++] |
H18O10Sr | |
MFCD00150395 | |
strontium(2+) octahydrate dihydroxide |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 242-367-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only