Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium hexafluoro-2,4-pentanedionate

$150.68 - $150.68

Chemical Identifiers

Chemical Name or Material Strontium hexafluoro-2,4-pentanedionate
CAS 121012-89-3
Formula Weight 501.75
MDL Number MFCD00064760
Synonym Strontium hexafluoroacetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1470503

  SDS
alfa aesar™
1470503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1470506

  SDS
alfa aesar™
1470506
5g Each for $150.68
Description & Specifications

Specifications

Formula Weight 501.75
Color White
Chemical Name or Material Strontium hexafluoro-2,4-pentanedionate
Physical Form Crystalline
MDL Number MFCD00064760
CAS 121012-89-3
Synonym Strontium hexafluoroacetylacetonate
Boiling Point 220°C (0.02mmHg) (subl.)
Melting Point ≈260°C (dec.)
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.