missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium fluoride, Optical Grade

Manufacturer:  Alfa Aesar™ 1102004

 View more versions of this product

Catalog No. AA1102004



Specifications

Specifications

Strontium fluoride
F2Sr
14,8842
FVRNDBHWWSPNOM-UHFFFAOYSA-L
strontium;difluoride
82210
Broken Crystals
4.240g/mL
1.442
2g
7783-48-4
MFCD00011251
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
[F-].[F-].[Sr+2]
125.617
125.62
Optical
Odorless
1473°C

Chemical Identifiers

7783-48-4
125.617
FVRNDBHWWSPNOM-UHFFFAOYSA-L
82210
[F-].[F-].[Sr+2]
F2Sr
MFCD00011251
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
strontium;difluoride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WK8925000

TSCA : TSCA

Documents