Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium fluoride, Optical Grade

Chemical Identifiers

Chemical Name or Material Strontium fluoride
CAS 7783-48-4
Purity Grade Optical Grade
Formula Weight 125.62
MDL Number MFCD00011251
Merck Index 14,8842
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1102004

  SDS
alfa aesar™
1102004
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1102009

  SDS
alfa aesar™
1102009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1102018

  SDS
alfa aesar™
1102018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 125.62
Odor Odorless
MDL Number MFCD00011251
Health Hazard 1 Warning
Purity Grade Optical Grade
Quantity 2g
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Strontium fluoride
CAS 7783-48-4
Refractive Index 1.442
Density 4.240g/mL
Physical Form Broken Crystals
RTECS Number WK8925000
Merck Index 14,8842
Melting Point 1473°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.