Learn More
Strontium fluoride, 99% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012201A1
Description
Strontium fluoride is used as an optical material for a small range of special applications. It also used as use is as a carrier of strontium-90 radioisotope in radioisotope thermoelectric generators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Strontium fluoride | |
7783-48-4 | |
Powder | |
F2Sr | |
14,8842 | |
Slightly soluble in water (11;7mg at 18°C/100mL), dilute acids; Decomposes in strong acids | |
[F-].[F-].[Sr++] | |
125.62 | |
125.62 | |
Odorless | |
4.240 g/mL |
1473°C | |
White | |
1 kg | |
MFCD00011251 | |
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g | |
FVRNDBHWWSPNOM-UHFFFAOYSA-L | |
strontium;difluoride | |
82210 | |
≤99% | |
1.442 |
Chemical Identifiers
7783-48-4 | |
125.62 | |
FVRNDBHWWSPNOM-UHFFFAOYSA-L | |
82210 | |
[F-].[F-].[Sr++] |
F2Sr | |
MFCD00011251 | |
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g | |
strontium;difluoride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 232-000-3
RTECSNumber : WK8925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only