Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium, distilled dendritic pieces, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Strontium
CAS 7440-24-6
Name Note Distilled dendritic pieces
MDL Number MFCD00134060
UN Number UN3208
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3862406

  SDS
alfa aesar™
3862406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging Ampule
Chemical Name or Material Strontium
TSCA TSCA
Name Note Distilled dendritic pieces
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00134060
CAS 7440-24-6
UN Number UN3208
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H261
In contact with water releases flammable gas.
Sensitivity Air & Moisture Sensitive
Health Hazard 3 GHS P Statement
P231+P232-P280-P233-P370+P378b-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: Special powder for metal fires.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
RTECS Number WK7700000
Quantity 5g

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P231+P232-P280-P233-P370+P378b-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: Special powder for metal fires.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H261
In contact with water releases flammable gas.

  • Ampouled under argon