Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium cyclohexanebutyrate, AAS

$25.08 - $87.43

Chemical Identifiers

Chemical Name or Material Strontium cyclohexanebutyrate
CAS 62638-05-5
Name Note AAS
Formula Weight 426.11
MDL Number MFCD00050727
Synonym Cyclohexanebutyric acid strontium salt; Strontium 4-cyclohexylburate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3973503

  SDS
alfa aesar™
3973503
1g Each for $25.08

AA3973506

  SDS
alfa aesar™
3973506
5g Each for $87.43
Description & Specifications

Specifications

Formula Weight 426.11
Chemical Name or Material Strontium cyclohexanebutyrate
TSCA TSCA
Name Note AAS
Physical Form Powder
MDL Number MFCD00050727
CAS 62638-05-5
Synonym Cyclohexanebutyric acid strontium salt; Strontium 4-cyclohexylburate
Quantity 1g