missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium chloride hexahydrate, Puratronic™, 99.9965% (metals basis)

Manufacturer:  Alfa Aesar™ 1087706

 View more versions of this product

Catalog No. AA1087706



Specifications

Specifications

Strontium chloride hexahydrate
(metals basis)
14,8840
Powder
Puratronic™
1.650
White
10025-70-4
MFCD00149865
266.62 (158.53 Anhydrous)
99.9965%
1.93g/mL
Hygroscopic
5g

Chemical Identifiers

10025-70-4
MFCD00149865
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

TSCA : TSCA

Documents