missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Strontium chloride hexahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 315085000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Strontium chloride hexahydrate | |
For analysis ACS | |
White | |
Crystals | |
99.0 to 103% (Complexometry) | |
SrCl2·6H2O | |
15, 8970 | |
AMGRXJSJSONEEG-UHFFFAOYSA-L | |
strontium(2+) hexahydrate dichloride | |
266.62 | |
ACS Reagent |
115.0°C | |
10025-70-4 | |
5.0 to 7.0 (5% soln. at 25°C) | |
500 g | |
Cl2H12O6Sr | |
MFCD00149865 | |
Solubility in water: 106.2 g/100mL (0°C). Other solubilities: soluble in alcohol | |
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] | |
266.61 | |
≥99% | |
Plastic bottle |
Chemical Identifiers
10025-70-4 | |
266.61 | |
AMGRXJSJSONEEG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] |
Cl2H12O6Sr | |
MFCD00149865 | |
strontium(2+) hexahydrate dichloride |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
RUO – Research Use Only