Please login to your online account to display your discounted pricing

Strontium Chloride, hexahydrate, 99.0 to 103.0%, ACS, EMD Millipore


Quantity 25kg

Manufacturer: emd millipore corporation  SX101520

 View more versions of this product

Catalog No. MSX101520

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Formula Weight 266.62
Color White
Quantity 25kg
Bulk Density 1050kg/m3
Density 1.95g/cm3 (20°C)
Molecular Formula Cl2Sr · 6H2O
CAS 10025-70-4
Assay Percent Range 99.0 to 103.0%
Name Note For Atomic Absorption Spectroscopy
Linear Formula SrCl2 · 6H2O
Recommended Storage Store without restrictions
Chemical Name or Material Strontium Chloride, hexahydrate

Specifications

Chemical Name or Material Strontium Chloride, hexahydrate
Name Note For Atomic Absorption Spectroscopy
CAS 10025-70-4
Physical Form Crystalline Solid
Assay Percent Range 99.0 to 103.0%
Molecular Formula Cl2Sr · 6H2O
Linear Formula SrCl2 · 6H2O
Formula Weight 266.62
Accessories

Store without restrictions