Please login to your online account to display your discounted pricing

Strontium Chloride, hexahydrate, 99.0 to 103.0%, ACS, EMD Millipore

$211.86 - $4,667.31

Chemical Identifiers

Chemical Name or Material Strontium Chloride, hexahydrate
Name Note For Atomic Absorption Spectroscopy
CAS 10025-70-4
Physical Form Crystalline Solid
Assay Percent Range 99.0 to 103.0%
Molecular Formula Cl2Sr · 6H2O
Linear Formula SrCl2 · 6H2O
Formula Weight 266.62
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MSX10151

 
emd millipore corporation
SX10151
500g Each for $211.86

MSX101520

 
emd millipore corporation
SX101520
25kg Each for $4,667.31
Description & Specifications

Specifications

Physical Form Crystalline Solid
Formula Weight 266.62
Color White
Quantity 500g
Bulk Density 1050kg/m3
Density 1.95g/cm3 (20°C)
Chemical Name or Material Strontium Chloride, hexahydrate
Name Note For Atomic Absorption Spectroscopy
Assay Percent Range 99.0 to 103.0%
Molecular Formula Cl2Sr · 6H2O
Linear Formula SrCl2 · 6H2O
CAS 10025-70-4
Recommended Storage Store without restrictions
Safety and Handling

Store without restrictions