missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium Chloride, Anhydrous, 99.5% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1679014

 View more versions of this product

Catalog No. AA1679014



Specifications

Specifications

Strontium Chloride
99.5%, metal basis
MFCD00011249
-40 Mesh Powder
Anhydrous
10476-85-4
Cl2Sr
158.53
3.052
25g

Chemical Identifiers

10476-85-4
Cl2Sr
MFCD00011249
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection.

Harmful if swallowed. Irritating to respiratory system and skin. Risk of serious damage to eyes.

EINECSNumber : 233-971-6

SDS Documents