missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium chloride, 99.99%, (trace metal basis), anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC369740050

 View more versions of this product

Catalog No. AC369740050



Description

Description

for ultra trace analysis
Specifications

Specifications

Strontium chloride
54.4 to 55.9% (Sr)
SrCl2
15, 8970
158.53
Crystalline Powder
Trace Metal Basis
Solubility in water: soluble.
1250.0°C
873.0°C
10476-85-4
Cl2Sr
MFCD00011249
Solubility in water: soluble
158.53
99.99%
Glass bottle
0.01% max
White
5g

Chemical Identifiers

10476-85-4
Cl2Sr
158.53
MFCD00011249
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 233-971-6

SDS Documents