Please login to your online account to display your discounted pricing

Strontium Chloride, 6-Hydrate, Low in Magnesium, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  403601

 View more versions of this product

Catalog No. 02004353

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 266.62
CAS 10025-70-4
Chemical Name or Material Strontium Chloride
Name Note 6-Hydrate, Low in Magnesium
Molecular Formula SrCl2·6H2O
Quantity 500g
Packaging Poly Bottle
Specific Gravity 1L = 1.96kg

Specifications

Chemical Name or Material Strontium Chloride
CAS 10025-70-4
Name Note 6-Hydrate, Low in Magnesium
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula SrCl2·6H2O
Formula Weight 266.62