missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium carbonate, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1434330

 View more versions of this product

Catalog No. AA1434330



Specifications

Specifications

Strontium carbonate, 99%, Ba 1%
99%
MFCD00011250
LEDMRZGFZIAGGB-UHFFFAOYSA-L
strontium;carbonate
15407
-325 Mesh, Typically 1 Micron Powder
1633-05-2
CO3Sr
strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
C(=O)([O-])[O-].[Sr+2]
147.628
147.63
250g

Chemical Identifiers

1633-05-2
147.628
LEDMRZGFZIAGGB-UHFFFAOYSA-L
15407
C(=O)([O-])[O-].[Sr+2]
CO3Sr
MFCD00011250
strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
strontium;carbonate
Safety and Handling

Safety and Handling

EINECSNumber : 216-643-7

Documents