Please login to your online account to display your discounted pricing

Strontium carbonate, 99.99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Strontium carbonate
CAS 1633-05-2
Name Note (metals basis)
Molecular Formula SrCO3
Formula Weight 147.63
MDL Number MFCD00011250
Merck Index 14,8838
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3579314

  SDS
alfa aesar
3579314
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.63
Quantity 25g
MDL Number MFCD00011250
Odor Odorless
Density 3.70
Name Note (metals basis)
Chemical Name or Material Strontium carbonate
Melting Point 1100°C (Dec.)
TSCA TSCA
Physical Form Powder
Assay 99.99%
CAS 1633-05-2
Merck Index 14,8838
Molecular Formula SrCO3