Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium bromide, ultra dry, 99.995% (metals basis)

Chemical Identifiers

Chemical Name or Material Strontium bromide
CAS 10476-81-0
Name Note Ultra dry
Linear Formula SrBr2
Formula Weight 247.44
MDL Number MFCD00049548
Merck Index 14,8837
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3571703

  SDS
alfa aesar™
3571703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3571706

  SDS
alfa aesar™
3571706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.44
Packaging Ampule
Chemical Name or Material Strontium bromide
TSCA TSCA
Odor Odorless
Name Note Ultra dry
Physical Form Powder
Assay 99.995%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Linear Formula SrBr2
MDL Number MFCD00049548
CAS 10476-81-0
Merck Index 14,8837
Color White
Melting Point 643°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Storage Note 1
  • Ampouled under argon
Density 4.216g/mL
RTECS Number WK8050000

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon