missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium bromide hexahydrate, 95%, Alfa Aesar™

Manufacturer:  Alfa Aesar 7210522

 View more versions of this product

Catalog No. AA7210522



Specifications

Specifications

Strontium bromide hexahydrate
95%
MFCD00150361
FLMJUJXBFKFYOZ-UHFFFAOYSA-L
strontium;dibromide;hexahydrate
165644
Crystalline
100g
7789-53-9
Br2H12O6Sr
unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
O.O.O.O.O.O.[Br-].[Br-].[Sr+2]
355.518
355.53 (247.44 Anhydrous)
2.386 g/mL

Chemical Identifiers

7789-53-9
355.518
FLMJUJXBFKFYOZ-UHFFFAOYSA-L
165644
O.O.O.O.O.O.[Br-].[Br-].[Sr+2]
Br2H12O6Sr
MFCD00150361
unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
strontium;dibromide;hexahydrate
Safety and Handling

Safety and Handling

EINECSNumber : 233-969-5

SDS Documents