missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium bromide, anhydrous, 99% (metals basis excluding Ba), Ba 0.1% max

Manufacturer:  Alfa Aesar™ 1679130

 View more versions of this product

Catalog No. AA1679130



Specifications

Specifications

Strontium bromide
10476-81-0
(metals basis excluding Ba), Ba 0.1% max.
SrBr2
14,8837
[Br-].[Br-].[Sr+2]
247.428
247.44
4.216g/mL
Hygroscopic
643°C
Anhydrous
99%
Br2Sr
MFCD00049548
YJPVTCSBVRMESK-UHFFFAOYSA-L
strontium;dibromide
5360351
Granular
Odorless
White
250g

Chemical Identifiers

10476-81-0
247.428
YJPVTCSBVRMESK-UHFFFAOYSA-L
strontium;dibromide
Br2Sr
MFCD00049548
5360351
[Br-].[Br-].[Sr+2]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

RTECSNumber : WK8050000

TSCA : TSCA

SDS Documents