Learn More
Strontium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04709906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Strontium bis(trifluoromethylsulfonyl)imide | |
White | |
Solid | |
C4F12N2O8S4Sr | |
MFCD23380176 | |
Bis(trifluoromethane)sulfonimide strontium salt; Bis(trifluoromethanesulfonyl)imide strontium salt | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sr+2] | |
647.891 | |
647.91 |
>200°C | |
862121-57-1 | |
5 g | |
[(CF3SO2)2N]2Sr | |
UN3261 | |
DIMYICIXZTVMOY-UHFFFAOYSA-N | |
strontium;bis(trifluoromethylsulfonyl)azanide | |
134159298 | |
Hygroscopic |
Chemical Identifiers
862121-57-1 | |
647.891 | |
DIMYICIXZTVMOY-UHFFFAOYSA-N | |
134159298 | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sr+2] |
C4F12N2O8S4Sr | |
MFCD23380176 | |
Bis(trifluoromethane)sulfonimide strontium salt; Bis(trifluoromethanesulfonyl)imide strontium salt | |
strontium;bis(trifluoromethylsulfonyl)azanide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only