missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium acetate hydrate, Puratronic™, 99.9965% (metals basis)

Manufacturer:  Alfa Aesar™ 1299014

 View more versions of this product

Catalog No. AA1299014



Specifications

Specifications

Strontium acetate hydrate
(metals basis)
Sr(OOCCH3)2·xH2O
3692534
strontium acetate, strontium diacetate, acetic acid, strontium salt, strontium di acetate, unii-4sl32ymy7b, 4sl32ymy7b, acetic acid, strontium salt 2:1, acmc-1bn7n, strontium 2+ ion diacetate, ksc492a5d
CC(=O)[O-].CC(=O)[O-].[Sr+2]
205.708
205.71 (Anhydrous)
99.9965%
Hygroscopic
25g
543-94-2
C4H6O4Sr
MFCD00036392
14,8836
RXSHXLOMRZJCLB-UHFFFAOYSA-L
strontium;diacetate
10987
Crystalline
Puratronic™
150°C (in -0.5H2O)

Chemical Identifiers

543-94-2
205.708
RXSHXLOMRZJCLB-UHFFFAOYSA-L
10987
CC(=O)[O-].CC(=O)[O-].[Sr+2]
C4H6O4Sr
MFCD00036392
strontium acetate, strontium diacetate, acetic acid, strontium salt, strontium di acetate, unii-4sl32ymy7b, 4sl32ymy7b, acetic acid, strontium salt 2:1, acmc-1bn7n, strontium 2+ ion diacetate, ksc492a5d
strontium;diacetate
Safety and Handling

Safety and Handling

RTECSNumber : AJ4725000

TSCA : TSCA

Documents