Learn More
Strontium acetate hemihydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012203A1
Description
Strontium acetate is used as a catalyst, as a chemical intermediate and in medicine. The primary application of strontium acetate is as an active component in toothpastes. This compound is used to desensitize teeth to extremes of hot and cold, sweets, acids, or touch.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Strontium acetate hemihydrate | |
14692-29-6 | |
1 kg | |
14,8836 | |
214.72 (205.71 Anhydrous) | |
2.099 |
150°C (in -0.5H2O) | |
Crystalline | |
MFCD00198064 | |
Soluble in water and slightly soluble in alcohol. | |
Reagent |
Chemical Identifiers
14692-29-6 | |
MFCD00198064 |
Safety and Handling
EINECSNumber : 208-854-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only