missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Strontium AA Standard, 1000 ppm Sr in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company ASR1KN500
Specifications
Strontium AA Standard | |
Natural Poly Bottle | |
94.0,4.22,0.23 | |
Colorless | |
N2O6Sr | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
strontium(2+) dinitrate | |
24848 |
10042-76-9,7697-37-2,7732-18-5 | |
500 mL | |
97.0,4.39,0.24 | |
1,000ppm Sr | |
MFCD00011248 | |
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
211.63 | |
1 mL = 1 mg Sr |
Chemical Identifiers
10042-76-9 | |
211.63 | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
strontium(2+) dinitrate |
N2O6Sr | |
MFCD00011248 | |
24848 | |
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |