Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium 2,4-pentanedionate hydrate

$30.74 - $271.22

Chemical Identifiers

Chemical Name or Material Strontium 2,4-pentanedionate hydrate
CAS 12193-47-4
Formula Weight 285.84 (Anhydrous)
MDL Number MFCD00013502
Synonym Strontium acetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1461406

  SDS
alfa aesar™
1461406
5g Each for $30.74

AA1461414

  SDS
alfa aesar™
1461414
25g Each for $97.49

AA1461422

  SDS
alfa aesar™
1461422
100g Each for $271.22
Description & Specifications

Specifications

Formula Weight 285.84 (Anhydrous)
Synonym Strontium acetylacetonate
Solubility Information Soluble in warm methoxyethanol, pyridine
Chemical Name or Material Strontium 2,4-pentanedionate hydrate
Physical Form Powder
Quantity 5g
CAS 12193-47-4
MDL Number MFCD00013502
Melting Point 200°C (dec.)