missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strong Acid Solution, for Phosphorus Analysis, (with extraction), Ricca Chemical

For Analysis of phosphorus

$100.48 - $100.48

Chemical Identifiers

CAS 7664-93-9 (Sulfuric Acid),7697-37-2 (Nitric Acid),7732-18-5 (Water, Deionized)
Physical Form Liquid
Chemical Name or Material Strong Acid Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
8080-16 Ricca Chemical Company
RI808016
Poly natural 500mL Each for $100.48
R80800001A Ricca Chemical Company
RIR80800001A
Plastic Bottle 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7664-93-9 (Sulfuric Acid),7697-37-2 (Nitric Acid),7732-18-5 (Water, Deionized)
Liquid
Strong Acid Solution

Specifications

Liquid
500mL
7697-37-2 (Nitric Acid),7732-18-5 (Water, Deionized),7664-93-9 (Sulfuric Acid)
Clear colorless
Poly natural
Strong Acid Solution