Please login to your online account to display your discounted pricing

Strong Acid Solution, for Phosphorus Analysis, (with extraction), Ricca Chemical


For Analysis of phosphorus

Quantity 1L
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR80800001A

 View more versions of this product

Catalog No. R80800001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Strong Acid Solution
CAS 7664-93-9 (Sulfuric Acid)
CAS 7697-37-2 (Nitric Acid)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
Packaging Plastic Bottle
Color Clear colorless
Quantity 1L

Specifications

Chemical Name or Material Strong Acid Solution
CAS 7664-93-9 (Sulfuric Acid)
CAS 7697-37-2 (Nitric Acid)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid