missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strong Acid Solution, For Phosphorus, LabChem™

$89.97 - $89.97

Chemical Identifiers

CAS 7732-18-5,7664-93-9,7697-37-2
CAS Max % 58.43,41.17,0.4
Linear Formula H2SO4
Formula Weight 98.08
UN Number UN2796
Chemical Name or Material Strong Acid Solution, For Phosphorus
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC253701 Lab Chem
LC253701
500mL Each for $89.97
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7664-93-9,7697-37-2
H2SO4
UN2796
58.43,41.17,0.4
98.08
Strong Acid Solution, For Phosphorus

Specifications

7732-18-5,7664-93-9,7697-37-2
Liquid
98.08
UN2796
Poly Bottle
1.31g/mL
2.1cSt
Clear, colorless
Strong Acid Solution, For Phosphorus
58.43,41.17,0.4
H2SO4
1.31g/mL
Passes Test
Soluble in water
Room Temperature
Sulfur compounds; Nitrogen oxides
500mL

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear eye protection, face protection, protective gloves, protective clothing.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.

Danger