missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile

Manufactured with optically clear polystyrene for accurate measurement. Corning™ Stripette™ Individually Wrapped Plastic or Paper/Plastic Disposable Polystyrene Serological Pipets are ideal for sterile tissue culture applications, especially while wearing latex gloves, as the wrap reduces static cling.

Manufacturer:  Corning™ C4489

 View more versions of this product

Catalog No. 07-200-575



Description

Description

  • Sterile
  • RNase-/DNase-free
  • Nonpyrogenic
  • Noncytotoxic
  • BSE/TSE-free
  • Exclusive anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Bidirectional graduations provide choice of ascending and descending scales
  • Negative graduations allow additional working volume
  • Three packaging options: Individually wrapped, paper/plastic; Individually wrapped, clear plastic; bulk packed
  • Volumetric accuracy of ±2% at full capacity
Specifications

Specifications

±2%
Yes
0.2 mL
10 mL
200/Cs.
±2%
34.4 cm
Sterile
25 mL
Non-pyrogenic, DNase, RNase-free
Yes
13.54 in.
Individually Wrapped, Paper/Plastic Packaging
Round
Red
Polystyrene
Serological Pipet
SDS
Documents
Product Certifications
Accessories