Please login to your online account to display your discounted pricing

Streptozocin, Tocris

DNA alkylator; antitumor and induces diabetes

$76.90 - $231.77

Chemical Identifiers

Physical Form Solid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

1621100

 
r & d systems
RDS1621100
Each for $76.90

1621500

 
r & d systems
RDS1621500
Each for $231.77
Description & Specifications

Specifications

Formula Weight 265.22
Quantity 100mg
Physical Form Solid
Chemical Name or Material Streptozocin
Molecular Formula C8H15N3O7
CAS 18883-66-4
Synonym 2-Deoxy-2-[[(methylnitrosoamino)carbonyl]amino]-D-glucose
Recommended Storage Store at -20°C
Assay Percent Range >95%
Safety and Handling

Store at -20°C