Please login to your online account to display your discounted pricing

eBiosciencetrade; Streptavidin eFluor™ 570

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

$131.95 - $323.35

Specifications

For Use With (Application) Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Immunocytochemistry
Synonym SA, Sav
Conjugate eFluor 570
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C. Do not freeze. Light-sensitive material.
View More Specs

 Alert

Contains Sodium azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123611

 
ebioscience
LS41431780
25μg Each for $131.95

501123612

 
ebioscience
LS41431782
100μg Each for $323.35
Description & Specifications

Specifications

For Use With (Application) Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Immunocytochemistry
Synonym SA, Sav
Conjugate eFluor 570
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C. Do not freeze. Light-sensitive material.