Learn More
Streamlight™ Rechargeable LiteBox™ Lanterns
Adjustable swivel neck allows for rotation of the lamp head
$282.03 - $298.10
Specifications
Battery Type | 12 AH |
---|---|
Certifications/Compliance | NFPA 1901-12-1.7 |
Product Type | Safety Lantern |
Height (English) | 7 in. |
Height (Metric) | 17.8 cm |
Catalog Number | Mfr. No. | Color | Style | Wattage | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Style | Wattage | Price | Quantity | |||
18-999-329
|
Streamlight™
45102 |
Orange | Swivel neck, Vehicle-Mount System | 20 W |
Each for $282.03
Case of 4 Each for $1,128.12 |
|
29-635-999
|
Streamlight™
45116 |
Orange | Swivel neck, Standard System | 8 W |
Each for $298.10
|
|
Description
- Light source: High-intensity halogen bi-pin bulb in 8 or 20w spot for 8 or 20w bulbs; 3.5 for 20w
- Light output: see Ordering Table
- 12AH battery provides up to nine hours of run time; battery is rechargeable up to 500 times
- LEDs provide battery status: red is charging, green is fully charged
- High-impact ABS thermoplastic construction sealed to resist water, sand and dust
- High-intensity sealed halogen beam
- Rubberized impact bumper and a protective lens ring
Standard System
Ideal for field maintenance, inspection of construction sites, and remote equipment; international safety orange or high-visibility yellow; includes shoulder strap; comes with AC and DC chargers and charge rack.
Vehicle Mount System
International safety orange or high-visibility yellow; comes with vehicle-mount hard-wire rack
and shoulder carrying strap.
Power Failure System
International safety orange or beige; comes with AC/DC chargers and “smart” power failure circuitry that automatically goes on when the power goes off.
Order Info
Dimensions: L × W × H: 11.5 × 5.1 × 7 in. (29.2 × 13 × 17.8cm).
Certifications
Most models meet applicable European Community Directives.
Compliance
Rack meets NFPA 1901-12-1.7 mounted in any position.
Warranty
Limited lifetime.
Specifications
12 AH | |
Safety Lantern | |
17.8 cm | |
29.2 cm | |
High-intensity halogen bi-pin bulb in 8 or 20 watt spot for 8- or 20 watt bulbs, 3.5 for 20 watt | |
7.4 lb. |
NFPA 1901-12-1.7 | |
7 in. | |
11.5 in. | |
LED | |
ABS Thermoplastic | |
3.3 kg |