Learn More
Streamlight™ Rechargeable LiteBox™ Lanterns
Adjustable swivel neck allows for rotation of the lamp head
Supplier: Streamlight™ 45116
Description
- Light source: High-intensity halogen bi-pin bulb in 8 or 20w spot for 8 or 20w bulbs; 3.5 for 20w
- Light output: see Ordering Table
- 12AH battery provides up to nine hours of run time; battery is rechargeable up to 500 times
- LEDs provide battery status: red is charging, green is fully charged
- High-impact ABS thermoplastic construction sealed to resist water, sand and dust
- High-intensity sealed halogen beam
- Rubberized impact bumper and a protective lens ring
Standard System
Ideal for field maintenance, inspection of construction sites, and remote equipment; international safety orange or high-visibility yellow; includes shoulder strap; comes with AC and DC chargers and charge rack.
Vehicle Mount System
International safety orange or high-visibility yellow; comes with vehicle-mount hard-wire rack
and shoulder carrying strap.
Power Failure System
International safety orange or beige; comes with AC/DC chargers and “smart” power failure circuitry that automatically goes on when the power goes off.
Order Info
Dimensions: L x W x H: 11.5 x 5.1 x 7 in. (29.2 x 13 x 17.8cm).
Certifications
Most models meet applicable European Community Directives.
Compliance
Rack meets NFPA 1901-12-1.7 mounted in any position.
Warranty
Limited lifetime.
Specifications
Safety Lantern | |
120 V | |
7.4 lb. | |
12 AH | |
Orange | |
Fire and Rescue, Industrial Safety, Utilities, Emergency Medical Services, Homeowners, Sporting Goods, Pest Control | |
17.8 cm | |
11.5 in. | |
150 lm |
Halogen | |
8 W | |
3.3 kg | |
NFPA 1901-12-1.7 | |
Swivel neck, Standard System | |
7 in. | |
Shoulder strap | |
29.2 cm | |
ABS Thermoplastic |