missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Straight-Tube Bunsen Burners
Noiseless burners allow separate control of both flame temperature and flame height.
Manufacturer: Fisher Science Education HUMH6225
Description
- Threaded-aluminum air-control sleeve
- Accurately machined, fine-pitch brass needle valve with internal stuffing box and self-sealing packing
- Quick adjustment provides a clear flame of any height between 0.5 to 12 in. (1.3 to 30.5cm)
- Completely rustproof
- Octagon-shaped, nickel-plated, die-cast base, 3.5 in. (8.9cm)
- Tapered and serrated gas inlet, 0.39 in. (1cm)
- Aluminum mixing tube
- Burners will produce in excess of 5000 BTU per hour at full valve opening
- Gas inlet/tubing size: 0.31 in. (11mm)
Specifications
Noiseless Burners Allow Separate Bontrol of both Flame Temperature and Flame Height | |
Bunsen Burner | |
2000 to 3200 BTU | |
6 in. | |
0.75 in. | |
0.44 in. | |
0.79 cm |
LPG | |
Noiseless burners allow separate control of both flame temperature and flame height | |
Aluminum Mixing Tube, Die-Cast Base, Octagon Shaped, Nickel-Plated Tube | |
15.2 cm | |
0.31 in. | |
1.9 cm | |
1.1 cm |